Learn More
(+/-)-Camphene, tech. (sum of camphene + fenchene)
CAS: 79-92-5 | C10H16 | 136.24 g/mol
$370.82
Chemical Identifiers
| CAS | 79-92-5 |
|---|---|
| Molecular Formula | C10H16 |
| Molecular Weight (g/mol) | 136.24 |
| MDL Number | MFCD00066603 |
| InChI Key | CRPUJAZIXJMDBK-UHFFFAOYNA-N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAA180400B
|
N/A | 1000 g |
Each for $370.82
|
|
|||||
Description
Camphene is used in the preparation of fragrances and as a food additive for flavoring.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCamphene is used in the preparation of fragrances and as a food additive for flavoring.
Solubility
Insoluble in water. Soluble in alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| 79-92-5 | |
| 0.85 | |
| 34°C (94°F) | |
| MFCD00066603 | |
| 14,1730 | |
| Insoluble in water. Soluble in alcohol. | |
| CC1(C2CCC(C2)C1=C)C | |
| 136.24 | |
| 136.24 | |
| (+/-)-Camphene, sum of camphene + fenchene |
| ∼35°C | |
| 159°C to 160°C | |
| C10H16 | |
| UN1325 | |
| 1s-2,2-dimethyl-3-methylidenebicyclo 2.2.1 heptane, s-camphene, 1s,4r-2,2-dimethyl-3-methylene-bicyclo 2.2.1 heptane | |
| CRPUJAZIXJMDBK-UHFFFAOYNA-N | |
| (1S,4S)-3,3-dimethyl-2-methylidenebicyclo[2.2.1]heptane | |
| 6560154 | |
| Technical |
Safety and Handling
GHS H Statement
H228-H319
Flammable solid.
Causes serious eye irritation.
P210-P240-P241-P264b-P280-P305+P351+P338-P370+P378q
H228-H319
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
RTECSNumber : EX1055000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only