missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Camphene, tech. (sum of camphene + fenchene)

Catalog No. AAA180400B
Change view
Click to view available options
Quantity:
1000 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA180400B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA180400B Supplier Thermo Scientific Chemicals Supplier No. A180400B
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 79-92-5 | C10H16 | 136.24 g/mol

Camphene is used in the preparation of fragrances and as a food additive for flavoring.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Camphene is used in the preparation of fragrances and as a food additive for flavoring.

Solubility
Insoluble in water. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 79-92-5
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00066603
InChI Key CRPUJAZIXJMDBK-UHFFFAOYNA-N

Specifications

Melting Point ∼35°C
Density 0.85
Boiling Point 159°C to 160°C
Flash Point 34°C (94°F)
Quantity 1000 g
UN Number UN1325
Merck Index 14,1730
Synonym 1s-2,2-dimethyl-3-methylidenebicyclo 2.2.1 heptane, s-camphene, 1s,4r-2,2-dimethyl-3-methylene-bicyclo 2.2.1 heptane
Solubility Information Insoluble in water. Soluble in alcohol.
SMILES CC1(C2CCC(C2)C1=C)C
IUPAC Name (1S,4S)-3,3-dimethyl-2-methylidenebicyclo[2.2.1]heptane
PubChem CID 6560154
Formula Weight 136.24
Grade Technical
Chemical Name or Material (+/-)-Camphene, sum of camphene + fenchene
Show More Show Less
Hazard Category H228-H319
Hazard Statement GHS H Statement
H228-H319
Flammable solid.
Causes serious eye irritation.
Precautionary Statement P210-P240-P241-P264b-P280-P305+P351+P338-P370+P378q
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
RTECSNumber EX1055000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only