missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+/-)-Camphene (contains ca. 20% Tricyclene) 78.0+%, TCI America™
Supplier: TCI America C0009100G
Specifications
| (+/-)-Camphene (contains ca. 20% Tricyclene) | |
| 46°C | |
| 159°C | |
| MFCD00066603 | |
| 1325 | |
| CRPUJAZIXJMDBK-UHFFFAOYSA-N | |
| 3,3-dimethyl-2-methylidenebicyclo[2.2.1]heptane | |
| 6616 | |
| 136.24 | |
| Crystalline Lumps |
| 79-92-5 | |
| White-Yellow | |
| C10H16 | |
| 100 g | |
| camphene, comphene, 2,2-dimethyl-3-methylenenorbornane, 3,3-dimethyl-2-methylenenorbornane, 3,3-dimethyl-2-methylenenorcamphane, +/--camphene, 2,2-dimethyl-3-methylenebicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylidenebicyclo 2.2.1 heptane, bicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylene, fema no. 2229 | |
| CC1(C2CCC(C2)C1=C)C | |
| 136.238 | |
| CHEBI:3830 | |
| ≥78.0% (GC) |
Chemical Identifiers
| 79-92-5 | |
| 136.238 | |
| CRPUJAZIXJMDBK-UHFFFAOYSA-N | |
| 6616 | |
| 3,3-dimethyl-2-methylidenebicyclo[2.2.1]heptane |
| C10H16 | |
| MFCD00066603 | |
| camphene, comphene, 2,2-dimethyl-3-methylenenorbornane, 3,3-dimethyl-2-methylenenorbornane, 3,3-dimethyl-2-methylenenorcamphane, +/--camphene, 2,2-dimethyl-3-methylenebicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylidenebicyclo 2.2.1 heptane, bicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylene, fema no. 2229 | |
| CHEBI:3830 | |
| CC1(C2CCC(C2)C1=C)C |
Safety and Handling
EINECSNumber : (4)-0613
RTECSNumber : EX1055000
TSCA : Yes