missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+/-)-Camphene (contains ca. 20% Tricyclene) 78.0+%, TCI America™
$32.28 - $39.87
Chemical Identifiers
| CAS | 79-92-5 |
|---|---|
| Molecular Formula | C10H16 |
| Molecular Weight (g/mol) | 136.238 |
| MDL Number | MFCD00066603 |
| InChI Key | CRPUJAZIXJMDBK-UHFFFAOYSA-N |
| Synonym | camphene, comphene, 2,2-dimethyl-3-methylenenorbornane, 3,3-dimethyl-2-methylenenorbornane, 3,3-dimethyl-2-methylenenorcamphane, +/--camphene, 2,2-dimethyl-3-methylenebicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylidenebicyclo 2.2.1 heptane, bicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylene, fema no. 2229 |
| PubChem CID | 6616 |
| ChEBI | CHEBI:3830 |
| IUPAC Name | 3,3-dimethyl-2-methylidenebicyclo[2.2.1]heptane |
| SMILES | CC1(C2CCC(C2)C1=C)C |
Chemical Identifiers
| C10H16 | |
| MFCD00066603 | |
| camphene, comphene, 2,2-dimethyl-3-methylenenorbornane, 3,3-dimethyl-2-methylenenorbornane, 3,3-dimethyl-2-methylenenorcamphane, +/--camphene, 2,2-dimethyl-3-methylenebicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylidenebicyclo 2.2.1 heptane, bicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylene, fema no. 2229 | |
| CHEBI:3830 | |
| CC1(C2CCC(C2)C1=C)C |
Specifications
| 79-92-5 | |
| White-Yellow | |
| C10H16 | |
| 1325 | |
| CRPUJAZIXJMDBK-UHFFFAOYSA-N | |
| 3,3-dimethyl-2-methylidenebicyclo[2.2.1]heptane | |
| 6616 | |
| 136.24 | |
| Crystalline Lumps |
| 46°C | |
| 159°C | |
| MFCD00066603 | |
| camphene, comphene, 2,2-dimethyl-3-methylenenorbornane, 3,3-dimethyl-2-methylenenorbornane, 3,3-dimethyl-2-methylenenorcamphane, +/--camphene, 2,2-dimethyl-3-methylenebicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylidenebicyclo 2.2.1 heptane, bicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylene, fema no. 2229 | |
| CC1(C2CCC(C2)C1=C)C | |
| 136.238 | |
| CHEBI:3830 | |
| ≥78.0% (GC) | |
| (+/-)-Camphene (contains ca. 20% Tricyclene) |
Safety and Handling
EINECSNumber : (4)-0613
RTECSNumber : EX1055000
TSCA : Yes