missing translation for 'onlineSavingsMsg'
Learn More

(-)-Camphanic Chloride 97.0+%, TCI America™

Catalog No. p-7124747
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10221G 1 g
C10225G 5 g
C102225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C10221G Supplier TCI America Supplier No. C10221G
Only null left

Chemical Identifiers

CAS 39637-74-6
Molecular Formula C10H13ClO3
Molecular Weight (g/mol) 216.661
MDL Number MFCD00135626
InChI Key PAXWODJTHKJQDZ-QVDQXJPCSA-N
Synonym 1s-4,7,7-trimethyl-3-oxo-2-oxabicyclo 2.2.1 heptane-1-carbonyl chloride, --1s,4r-camphanoyl chloride, 1s---camphanic acid chloride
PubChem CID 12364938
IUPAC Name (4S)-1,7,7-trimethyl-2-oxo-3-oxabicyclo[2.2.1]heptane-4-carbonyl chloride
SMILES CC1(C2(CCC1(OC2=O)C(=O)Cl)C)C

Specifications

Melting Point 70°C
Color White-Yellow
Quantity 1 g
UN Number 3261
Formula Weight 216.66
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Camphanic Chloride
TSCA No
Recommended Storage Refrigerator