missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CaMKII Pan Recombinant Rabbit Monoclonal Antibody (23GB4370)
GREENER_CHOICE

Catalog No. MA552702
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552702 100 μL
MA552703 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552702 Supplier Invitrogen™ Supplier No. MA552702
Only null left

Rabbit Recombinant Monoclonal Antibody

CaMKII Pan Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

The Ca2+/calmodulin-dependent protein kinases (CaM kinases) comprise a structurally related subfamily of serine/threonine kinases which include CaMKI, CaMKII and CaMKIV. CaMKII is an ubiquitously expressed serine/threonine protein kinase that is activated by Ca2+/calmodulin (CaM) binding and autophosphorylation and has been implicated in regulation of the cell cycle and transcription. It is involved in dendritic spine and synapse formation, neuronal plasticity and regulation of sarcoplasmic reticulum Ca2+ transport in skeletal muscle. In neurons, plays an essential structural role in the reorganization of the actin cytoskeleton during plasticity by binding and bundling actin filaments in a kinase-independent manner.

Specifications

Antigen CaMKII Pan
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4370
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Camk2b
Gene Accession No. P08413, P11275, P11730, P11798, P15791, P28652, Q13554, Q13555, Q13557, Q6PHZ2, Q923T9, Q9UQM7
Gene Alias [d]-CaMKII; 2810011D23Rik; 8030469K03Rik; alpha CaM kinase II; alpha-CaMKII; Ca++/calmodulin-dependent protein kinase 2 delta subunit; Ca++/calmodulin-dependent protein kinase II beta subunit; Ca++/calmodulin-dependent protein kinase II delta subunit; Ca++/calmodulin-dependent protein kinase II, delta subunit; Ca2+/calmodulin-dependent protein kinase II; Ca2+/calmodulin-dependent protein kinase II alpha; calcium/calmodulin dependent protein kinase II alpha; calcium/calmodulin dependent protein kinase II beta; calcium/calmodulin dependent protein kinase II delta; calcium/calmodulin dependent protein kinase II gamma; calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha; calcium/calmodulin-dependent protein kinase (CaM kinase) II beta; calcium/calmodulin-dependent protein kinase (CaM kinase) II delta; calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma; calcium/calmodulin-dependent protein kinase II alpha; calcium/calmodulin-dependent protein kinase II alpha subunit; calcium/calmodulin-dependent protein kinase II alpha-B subunit; calcium/calmodulin-dependent protein kinase II beta; calcium/calmodulin-dependent protein kinase II delta; calcium/calmodulin-dependent protein kinase II gamma; calcium/calmodulin-dependent protein kinase II, beta; calcium/calmodulin-dependent protein kinase II, delta; calcium/calmodulin-dependent protein kinase type II alpha chain; calcium/calmodulin-dependent protein kinase type II beta chain; calcium/calmodulin-dependent protein kinase type II delta chain; calcium/calmodulin-dependent protein kinase type II gamma chain; calcium/calmodulin-dependent protein kinase type II subunit alpha; Calcium/calmodulin-dependent protein kinase type II subunit beta; calcium/calmodulin-dependent protein kinase type II subunit delta; Calcium/calmodulin-dependent protein kinase type II subunit gamma; CaM kinase II alpha subunit; CaM kinase II beta subunit; CaM kinase II delta subunit; caM kinase II subunit alpha; CaM kinase II subunit beta; caM kinase II subunit delta; CaM kinase II subunit gamma; CAM2; CAMK; CaMK II; CAMK1; Camk2; Camk2a; Camk2b; Camk2d; Camk2g; CAMKA; CAMKB; CAMKD; CAMKG; Camki; CaMKII; CAMK-II; CaMK-II alpha subunit; CaMK-II delta subunit; caMK-II subunit alpha; caMK-II subunit beta; caMK-II subunit delta; caMK-II subunit gamma; CaMKIINalpha; CaM-kinase II alpha chain; CaM-kinase II beta chain; CaM-kinase II delta chain; caM-kinase II gamma chain; Ck2b; KIAA0968; Kiaa4163; mKIAA0968; PK2CDD; PKCCD; proline rich calmodulin-dependent protein kinase; R74975; RATCAMKI
Gene Symbols Camk2a, Camk2b, CAMK2D, Camk2g
Host Species Rabbit
Immunogen A synthesized peptide derived from human CaMKII (550-666AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108058, 12322, 12323, 12325, 171140, 24245, 24246, 25400, 815, 816, 817, 818
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less