missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CaMKI gamma Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113240
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113240 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113240 Supplier Invitrogen™ Supplier No. PA5113240

Rabbit Polyclonal Antibody

CaMKI gamma Polyclonal Antibody for Western Blot, IHC (P), ELISA

CAMK1G serine/threonine kinase is a member of CAMK family and is a novel membrane-anchored neuronal Ca2+ calmodulin-dependent protein kinase. Full activation of CAMK1G requires both Ca2+ and calmodulin and phosphorylation by CAMKK. CAMK1G transcripts are most abundant in neurons, with the highest levels in limited nuclei such as the central nucleus of the amygdala (CeA) and the ventromedial hypothalamus.

Specifications

Antigen CaMKI gamma
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Camk1g
Gene Accession No. Q96NX5
Gene Alias calcium/calmodulin dependent protein kinase IG; calcium/calmodulin-dependent protein kinase I gamma; calcium/calmodulin-dependent protein kinase IG; calcium/calmodulin-dependent protein kinase type 1G; CaM kinase 1G; CaM kinase I gamma; caM kinase IG; CAMK1G; caMKI gamma; caM-KI gamma; caMKIG; CaMKIgamma; caMKI-gamma; caMK-like CREB kinase III; CLICK III; CLICK3; CLICKIII; CLICK-III; dJ272L16.1; RP1-272L16.2; VWS1
Gene Symbols Camk1g
Host Species Rabbit
Immunogen Recombinant Human Calcium/calmodulin-dependent protein kinase type 1G protein (217-476AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57172
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less