missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CaMKI gamma Polyclonal Antibody

Description
Recommended positive controls: U87-MG, SK-N-SH, IMR32, SK-N-AS, 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji, Mouse brain, Rat brain. Predicted reactivity: Mouse (98%), Rat (98%), Pig (96%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CAMK1G serine/threonine kinase is a member of CAMK family and is a novel membrane-anchored neuronal Ca2+ calmodulin-dependent protein kinase. Full activation of CAMK1G requires both Ca2+ and calmodulin and phosphorylation by CAMKK. CAMK1G transcripts are most abundant in neurons, with the highest levels in limited nuclei such as the central nucleus of the amygdala (CeA) and the ventromedial hypothalamus.
Specifications
Specifications
| Antigen | CaMKI gamma |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.84 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | Camk1g |
| Gene Accession No. | Q7TNJ7, Q91VB2, Q96NX5 |
| Gene Alias | calcium/calmodulin dependent protein kinase IG; calcium/calmodulin-dependent protein kinase I gamma; calcium/calmodulin-dependent protein kinase IG; calcium/calmodulin-dependent protein kinase type 1G; CaM kinase 1G; CaM kinase I gamma; caM kinase IG; CAMK1G; caMKI gamma; caM-KI gamma; caMKIG; CaMKIgamma; caMKI-gamma; caMK-like CREB kinase III; CLICK III; CLICK3; CLICKIII; CLICK-III; dJ272L16.1; RP1-272L16.2; VWS1 |
| Gene Symbols | Camk1g |
| Show More |