missing translation for 'onlineSavingsMsg'
Learn More

CaMKI delta Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501563166
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501563166 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501563166 Supplier Bethyl Laboratories Inc Supplier No. A302667A
Only null left

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Calcium/clamodulin-dependent protein kinase 1D-beta isoform is a novel calmodulin-kinase that exhibits Ca (2+)/CaM-dependent activity. CAM-Kkalpha upregulates CAMK1D-b activity through phosphorylation of its Thr180.

Specifications

Antigen CaMKI delta
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene CAMK1D
Gene Accession No. Q8IU85
Gene Alias CAMKI, MGC120317, MGC120318
Gene Symbols CAMK1D
Host Species Rabbit
Immunogen Between 335 and 385
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57118
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less