Learn More
Invitrogen™ Human CAMK2A (CaMKII Alpha), His Tag Recombinant Protein
Recombinant Protein
$908.00 - $12900.00
Specifications
| Accession Number | Q9UQM7 |
|---|---|
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 20mM tris with 50% glycerol, 100mM NaCl, 0.5mM EDTA, 2mM DTT, 0.05% NP-40 and no preservative; pH 7.5 |
| Gene ID (Entrez) | 815 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PR4586A
|
Invitrogen™
PR4586A |
1 mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
PV3142
|
Invitrogen™
PV3142 |
10 μg |
Each for $908.00
|
|
|||||
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length.
CAMK2A (CaMKII alpha) is a serine/threonine kinase that regulates a number of cellular functions including neuronal processes, gene expression, carbohydrate metabolism, cytoskeletal function, and calcium homeostatsis. Autophosphorylation of threonine 286 in the presence of Ca2+ and calmodulin activates CAMK2A with the ability to switch from calmodulin-dependent to a calmodulin independent activity. CaMKII is an ubiquitously expressed serine/threonine protein kinase that is activated by Ca2+/calmodulin (CaM) binding and autophosphorylation and has been implicated in regulation of the cell cycle and transcription. It is involved in dendritic spine and synapse formation, neuronal plasticity and regulation of sarcoplasmic reticulum Ca2+ transport in skeletal muscle. In neurons, plays an essential structural role in the reorganization of the actin cytoskeleton during plasticity by binding and bundling actin filaments in a kinase-independent manner.Specifications
| Q9UQM7 | |
| Kinase Assay | |
| 815 | |
| Human CAMK2A (CaMKII Alpha), His Tag | |
| Purified | |
| RUO | |
| CaMKII alpha | |
| CAMK2 Kinase Family | |
| Unconjugated | |
| Recombinant | |
| Full length | |
| Insect cells | |
| Liquid | |
| Signal Transduction |
| See Label | |
| 20mM tris with 50% glycerol, 100mM NaCl, 0.5mM EDTA, 2mM DTT, 0.05% NP-40 and no preservative; pH 7.5 | |
| 56.8 kDa | |
| 7.5 | |
| -80°C, Avoid Freeze/Thaw Cycles | |
| alpha CaM kinase II; alpha-CaMKII; Ca2+/calmodulin-dependent protein kinase II; Ca2+/calmodulin-dependent protein kinase II alpha; Calci; calcium/calmodulin dependent protein kinase II alpha; calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha; calcium/calmodulin-dependent protein kinase II alpha; calcium/calmodulin-dependent protein kinase II alpha subunit; calcium/calmodulin-dependent protein kinase II alpha-B subunit; calcium/calmodulin-dependent protein kinase type II alpha chain; calcium/calmodulin-dependent protein kinase type II subunit alpha; CaM kinase II alpha subunit; caM kinase II subunit alpha; CaMK II; CAMK2; Camk2a; CAMKA; CaMKII; CaMK-II alpha subunit; caMK-II subunit alpha; CaMKIINalpha; CaM-kinase II alpha chain; EC 2.7.11.17; KIAA0968; mKIAA0968; OTTHUMP00000165787; OTTHUMP00000165788; Phospho-CaMKI; PK2CDD; PKCCD; R74975 | |
| CAMK2A | |
| Protein | |
| Human | |
| His-tag | |
| Dry Ice | |
| Recombinant, Full Length | |
| Serine/Threonine Kinases |