Learn More
Invitrogen™ Human CAMK1D (CaMKI Delta), His Tag Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ PV3663
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.
CAMK1D (CaMK1 delta) is a serine/threonine kinase that is a member of the calcium/calmodulin-dependent protein kinase family. CAMK1D is expressed in polymorphonuclear leukocytes and may be part of the chemokine signal transduction pathway that regulates granulocyte function. It may also be involved in modulation of neuronal apoptosis. Calcium/clamodulin-dependent protein kinase 1D-beta isoform is a novel calmodulin-kinase that exhibits Ca (2+)/CaM-dependent activity. CAM-Kkalpha upregulates CAMK1D-b activity through phosphorylation of its Thr180.Specifications
| Q8IU85 | |
| Kinase Assay | |
| 57118 | |
| Human CAMK1D (CaMKI Delta), His Tag | |
| Purified | |
| -80°C, Avoid Freeze/Thaw Cycles | |
| A630059D12Rik; calcium/calmodulin dependent protein kinase ID; calcium/calmodulin-dependent protein kinase 1D; calcium/calmodulin-dependent protein kinase ID; calcium/calmodulin-dependent protein kinase type 1D; CaM kinase I delta; caM kinase ID; CaM-K1; CAMK1D; CAMKI; caMKI delta; caM-KI delta; CAMKID; CaMKIdelta; CaMKI-delta; camKI-like protein kinase; CKLiK; E030025C11Rik; mCKLiK; MGC120317; MGC120318; OTTHUMP00000019119; OTTHUMP00000019120; OTTHUMP00000045030; RGD1560691; RP11-462F15.1 | |
| CAMK1D | |
| Protein | |
| Human | |
| His-tag | |
| -80°C, Avoid Freeze/Thaw Cycles | |
| SerâThr Kinases: CAMK Group | |
| LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
| calciumâcalmodulin-dependent protein kinase ID | |
| Kinases & Inhibitors | |
| Liquid | |
| Signal Transduction |
| See Label | |
| 50mM tris with 0.02% Triton X-100, 150mM NaCl, 0.5mM EDTA, 2mM DTT, 50% glycerol and no preservative; pH 7.5 | |
| 47 kDa | |
| 7.5 | |
| 10 μg | |
| RUO | |
| CAMK1D | |
| CAMK1 Kinase Family | |
| Unconjugated | |
| Recombinant | |
| Full length | |
| Biochemical terbium phospho-specific activity, Biochemical binding/kinetic | |
| Dry Ice | |
| CAMK1D | |
| Insect cells | |
| Recombinant, Full Length | |
| Serine/Threonine Kinases |