missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calreticulin Recombinant Rabbit Monoclonal Antibody (23GB700)

Description
Calreticulin Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
Calr or Calreticulin is a calcium-binding chaperone that promotes folding, oligomeric assembly and quality control in the endoplasmic reticulum (ER) via the calreticulin/calnexin cycle. Calr, a lectin, interacts transiently with almost all of the monoglucosylated glycoproteins that are synthesized in the ER. Calr also interacts with with the DNA-binding domain of NR3C1 and mediates its nuclear export. It may also be involved in maternal gene expression regulation and oocyte maturation via the regulation of calcium homeostasis.
Specifications
Specifications
| Antigen | Calreticulin |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 23GB700 |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | CALR |
| Gene Accession No. | P14211, P18418, P27797 |
| Gene Alias | Autoantigen RO; CABP3; CALBP; calcium-binding protein 3; CALR; calrectulin; calregulin; Calreticulin; calreticulin precursor; cC1qR; CRP55; CRT; CRTC; Endoplasmic reticulum resident protein 60; epididymis secretory sperm binding protein Li 99n; ERp60; FLJ26680; grp60; HACBP; HEL-S-99n; I79_008346; RO; Ro/SS-A Antigen; Sicca syndrome antigen A (autoantigen Ro; calreticulin); SSA |
| Show More |