missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calreticulin Polyclonal Antibody, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt.
Calr or Calreticulin is a calcium-binding chaperone that promotes folding, oligomeric assembly and quality control in the endoplasmic reticulum (ER) via the calreticulin/calnexin cycle. Calr, a lectin, interacts transiently with almost all of the monoglucosylated glycoproteins that are synthesized in the ER. Calr also interacts with with the DNA-binding domain of NR3C1 and mediates its nuclear export. It may also be involved in maternal gene expression regulation and oocyte maturation via the regulation of calcium homeostasis.
Specifications
Specifications
| Antigen | Calreticulin |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.1% BSA and 0.09% sodium azide |
| Gene | CALR |
| Gene Accession No. | P14211, P27797 |
| Gene Alias | Autoantigen RO antibody, CALR antibody. |
| Gene Symbols | Calr |
| Host Species | Rabbit |
| Show More |