missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Calponin 2 Monoclonal Antibody (CP-93)
GREENER_CHOICE

Catalog No. PIMA549306
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549306 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549306 Supplier Invitrogen™ Supplier No. MA549306
Only null left

Mouse Monoclonal Antibody

Adding 1 mL of PBS buffer will yield a concentration of 100 μg/mL. Positive Control - WB: chicken heart tissue, chicken heart tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

The protein encoded by this gene, which can bind actin, calmodulin, troponin C, and tropomyosin, may function in the structural organization of actin filaments. The encoded protein could play a role in smooth muscle contraction and cell adhesion. Two transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen Calponin 2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone CP-93
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites, 1.2% sodium acetate with 2mg BSA and 0.01mg sodium azide
Gene Cnn2
Gene Accession No. Q99439
Gene Alias AA408047; AI324678; Calpo2; calponin 2; calponin H2, smooth muscle; Calponin-2; CLP2; CNN2; h2-calponin; LOW QUALITY PROTEIN: calponin-2; neutral calponin
Gene Symbols Cnn2
Host Species Mouse
Immunogen Turkey gizzard calponin.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100216000, 1265
Target Species Avian, Chicken, Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less