Learn More
OHAUS CalPak Certified Calibration Weight Set, ASTM Class 1
ASTM Class 1 Certified Calibration Weight Set with an Accredited Certificate
Supplier: OHAUS 80859549
Description
- Calibration Weight Sets Adjusted to Meet or Exceed ASTM Class 1-4 Tolerances
- Troemner ASTM Class 1-4 offer high accuracy with tight tolerances and uncertainties of less than 0.25 of the tolerance
- Weight surfaces meet or exceed ASTM and have a highly polished finish
- Available in high grade stainless steels including Troemner Alloy 8 which possesses closely controlled density, extremely low magnetic properties, good stability, and resistance to corrosion
- Available with certificate options traceable to NIST, including ISO/IEC 17025 accredited certification or traceable certification to aid in meeting SOP and regulatory requirements
Specifications
| Calibration Weight | |
| Stainless Steel | |
| 200g-10g | |
| Uncertainties of less than 1/4 of the tolerance. |