Learn More
OHAUS CalPak Certified Calibration Weight Set, ASTM Class 1
Catalog No. 11120743
Change view
Click to view available options
Weight (Metric):
100g-5g
1kg-50g
200g-10g
200g-2g
2kg-100g
2kg-20g
500g-20g
50g-2g
5kg-200g
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (Metric)
11120743 50g-2g
11120744 100g-5g
11120742 200g-2g
11120745 200g-10g
11120747 500g-20g
11120748 1kg-50g
11120741 2kg-20g
11120749 2kg-100g
11120746 5kg-200g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 11120743 Supplier OHAUS Supplier No. OHS30391795
Only null left
ASTM Class 1 Certified Calibration Weight Set with an Accredited Certificate
  • Calibration Weight Sets Adjusted to Meet or Exceed ASTM Class 1-4 Tolerances
  • Troemner ASTM Class 1-4 offer high accuracy with tight tolerances and uncertainties of less than 0.25 of the tolerance
  • Weight surfaces meet or exceed ASTM and have a highly polished finish
  • Available in high grade stainless steels including Troemner Alloy 8 which possesses closely controlled density, extremely low magnetic properties, good stability, and resistance to corrosion
  • Available with certificate options traceable to NIST, including ISO/IEC 17025 accredited certification or traceable certification to aid in meeting SOP and regulatory requirements
Specifications
Product Type Calibration Weight
Material Stainless Steel
Weight (Metric) 50g-2g
Tolerance Uncertainties of less than 1/4 of the tolerance.