Learn More
OHAUS CalPak Certified Calibration Weight Set, ASTM Class 1
ASTM Class 1 Certified Calibration Weight Set with an Accredited Certificate
Supplier:  OHAUS OHS30391790
 View more versions of this product
Catalog No. 11-120-742
Only null left
Explore available promotions
Description
Description
  • Calibration Weight Sets Adjusted to Meet or Exceed ASTM Class 1-4 Tolerances
  • Troemner ASTM Class 1-4 offer high accuracy with tight tolerances and uncertainties of less than 0.25 of the tolerance
  • Weight surfaces meet or exceed ASTM and have a highly polished finish
  • Available in high grade stainless steels including Troemner Alloy 8 which possesses closely controlled density, extremely low magnetic properties, good stability, and resistance to corrosion
  • Available with certificate options traceable to NIST, including ISO/IEC 17025 accredited certification or traceable certification to aid in meeting SOP and regulatory requirements
Specifications
Specifications
Calibration Weight
Stainless Steel
200g-2g
Uncertainties of less than 1/4 of the tolerance.
Product Suggestions
Product Suggestions