missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Calpain S1/S2 Monoclonal Antibody (28F3)
GREENER_CHOICE

Catalog No. pima134966
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA134966 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA134966 Supplier Invitrogen™ Supplier No. MA134966
Only null left

Mouse Monoclonal Antibody

This antibody specifically recognizes the 28 to 30 kD small subunit of Calpain 1 and Calpain 2 (mu calpain and m calpain). It recognizes both native and denatured protein in ELISA, Western Blot and Immunoprecipitation.

The calpains are calcium-dependent cysteine proteases that are widely expressed in mammalian systems. Both m-calpain (calpain II) and m-calpain (calpain I) are composed of an 80 kD subunit and a 30 kD subunit. Whereas the 30 kDa subunit is shared by both enzymes, the larger catalytic subunits are different and exhibit the distinct Ca++ requirements that are suggested by their names. Whereas m-calpain requires millimolar (mM) levels of calcium, m-calpain is active at micromolar (mM) concentrations of Ca++. In addition to the ubiquitously expressed m- and m- calpains, some tissue-specific calpains have been identified. The calpains appear to serve multiple physiological roles, and ideas concerning the functions of these enzymes are in a state of rapid flux.

Specifications

Antigen Calpain S1/S2
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 28F3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 3mM sodium azide; pH 7.5
Gene CAPNS1
Gene Accession No. P04632, P13135, Q96L46
Gene Alias 2310005G05Rik; 30K-2; calcium-activated neutral proteinase 2; Calcium-activated neutral proteinase small subunit; Calcium-dependent protease small subunit; Calcium-dependent protease small subunit 1; calcium-dependent protease small subunit 2; calcium-dependent protease, small subunit; calpain 2; calpain 2, (m/II) large subunit; calpain 2, large [catalytic] subunit; calpain 2, large subunit; calpain 4; calpain 4, small subunit (30K); calpain II regulatory subunit; calpain II regulatory subunit (EC 3.4.22.17); calpain large polypeptide L2; Calpain M-type; calpain regulatory subunit; Calpain small subunit 1; calpain small subunit 2; calpain, large polypeptide L2; calpain, small polypeptide; calpain, small subunit 1; calpain, small subunit 2; calpain-2 catalytic subunit; Calpain-2 large subunit; CALPAIN4; CANP; CANP 2; CANP small subunit; CANP2; CANPL2; CANPml; CANPS; Capa4; Capa-4; CAPN2; Capn4; CAPNS; CAPNS1; Capns2; CDPS; CSS1; CSS2; D7Ertd146e; M-calpain; mCANP; Millimolar-calpain
Gene Symbols CAPNS1, CAPNS2
Host Species Mouse
Immunogen Full length protein (Human). Antibody specifically recognizes small subunit.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100300424, 281664, 826, 84290
Target Species Human, Bovine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less