missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Calpain S1 Recombinant Rabbit Monoclonal Antibody (23GB3720)

Catalog No. MA553467
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553467 20 μL
MA553466 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553467 Supplier Invitrogen™ Supplier No. MA553467
Only null left

Rabbit Recombinant Monoclonal Antibody

Calpain S1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Calpains are a ubiquitous, well-conserved family of calcium-dependent, cysteine proteases. Calpain families have been implicated in neurodegenerative processes, as their activation can be triggered by calcium influx and oxidative stress. Calpain I and II are heterodimeric with distinct large subunits associated with common small subunits, all of which are encoded by different genes. Calpain S1 is a small subunit common to both calpain I and II and is associated with myotonic dystrophy.

Specifications

Antigen Calpain S1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3720
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CAPNS1
Gene Accession No. P04632
Gene Alias 30K; Calcium-activated neutral proteinase small subunit; calcium-dependent protease; Calcium-dependent protease small subunit; Calcium-dependent protease small subunit 1; calcium-dependent protease, small subunit; calpain 4; calpain 4, small subunit (30K); calpain II regulatory subunit; calpain II regulatory subunit (EC 3.4.22.17); calpain regulatory subunit; Calpain small subunit 1; calpain, small polypeptide; calpain, small subunit 1; CALPAIN4; CANP; CANP small subunit; CANPS; Capa4; Capa-4; Capn4; CAPNS; CAPNS1; CDPS; CSS1; D7Ertd146e; small pol; unknown protein
Gene Symbols CAPNS1
Host Species Rabbit
Immunogen A synthetic peptide of human Calpain small subunit 1 (50-100AA).
Purification Method Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 826
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less