missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Calmagite, pure, indicator grade

CAS: 3147-14-6 | C17H14N2O5S | 358.368 g/mol

$88.33 - $800.85

Chemical Identifiers

CAS 3147-14-6
Molecular Formula C17H14N2O5S
Molecular Weight (g/mol) 358.368
MDL Number MFCD00011656
InChI Key ASFVMSDYPYMUFL-UHFFFAOYSA-N
Synonym 3-Hydroxy-4-(2-hydroxy-5-methylphenylazo)-1-naphthalenesulfonic acid
PubChem CID 6364506
IUPAC Name 4-[(2-hydroxy-5-methylphenyl)hydrazinylidene]-3-oxonaphthalene-1-sulfonic acid
SMILES CC1=CC(=C(C=C1)O)NN=C2C3=CC=CC=C3C(=CC2=O)S(=O)(=O)O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC108170050
SDS View Documents
Thermo Scientific Chemicals
108170050
5 g Glass bottle
Each for $88.33
Only null left
 
AC108170100
SDS View Documents
Thermo Scientific Chemicals
108170100
10 g Glass bottle
Each for $133.12
Only null left
 
AC108170250
SDS View Documents
Thermo Scientific Chemicals
108170250
25 g Glass bottle
Each for $250.36
Only null left
 
AC108171000
SDS View Documents
Thermo Scientific Chemicals
108171000
100 g Glass bottle
Each for $800.85
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

3147-14-6
358.368
ASFVMSDYPYMUFL-UHFFFAOYSA-N
6364506
CC1=CC(=C(C=C1)O)NN=C2C3=CC=CC=C3C(=CC2=O)S(=O)(=O)O
C17H14N2O5S
MFCD00011656
3-Hydroxy-4-(2-hydroxy-5-methylphenylazo)-1-naphthalenesulfonic acid
4-[(2-hydroxy-5-methylphenyl)hydrazinylidene]-3-oxonaphthalene-1-sulfonic acid

Specifications

300.0°C
C17H14N2O5S
15, 1720
Solubility in water: soluble.
Authentic
4-[(2-hydroxy-5-methylphenyl)hydrazinylidene]-3-oxonaphthalene-1-sulfonic acid
6364506
Pure
300 min. (610nm, in buffer)
Black
Crystalline Powder
3147-14-6
MFCD00011656
3-Hydroxy-4-(2-hydroxy-5-methylphenylazo)-1-naphthalenesulfonic acid
ASFVMSDYPYMUFL-UHFFFAOYSA-N
CC1=CC(=C(C=C1)O)NN=C2C3=CC=CC=C3C(=CC2=O)S(=O)(=O)O
358.368
358.38
Glass bottle
suitable for determination of calcium and magnesium
5 g
Calmagite
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
Documents

Documents

RUO – Research Use Only