missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Calmagite, pure, indicator grade

Catalog No. AC108170050
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108170050 5 g Glass bottle
AC108170100 10 g Glass bottle
AC108170250 25 g Glass bottle
AC108171000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC108170050 Supplier Thermo Scientific Chemicals Supplier No. 108170050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3147-14-6 | C17H14N2O5S | 358.368 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 3147-14-6
Molecular Formula C17H14N2O5S
Molecular Weight (g/mol) 358.368
MDL Number MFCD00011656
InChI Key ASFVMSDYPYMUFL-UHFFFAOYSA-N
Synonym 3-Hydroxy-4-(2-hydroxy-5-methylphenylazo)-1-naphthalenesulfonic acid
PubChem CID 6364506
IUPAC Name 4-[(2-hydroxy-5-methylphenyl)hydrazinylidene]-3-oxonaphthalene-1-sulfonic acid
SMILES CC1=CC(=C(C=C1)O)NN=C2C3=CC=CC=C3C(=CC2=O)S(=O)(=O)O

Specifications

Melting Point 300.0°C
Merck Index 15, 1720
Solubility Information Solubility in water: soluble.
Infrared Spectrum Authentic
Formula Weight 358.38
Grade Pure
Packaging Glass bottle
Specific Extinction 300 min. (610nm, in buffer)
Suitability suitable for determination of calcium and magnesium
Color Black
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Calmagite
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only