missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calmagite, ACS, Spectrum™ Chemical
$416.69 - $20922.44
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18609551
|
Spectrum Chemical Mfg Cor
C23625GM |
5 g | Amber Glass Bottle |
Each for $416.69
|
|
||||
|
18609550
|
Spectrum Chemical Mfg Cor
C236225GM |
25 g | Amber Glass Bottle |
Each for $915.47
|
|
||||
|
18609549
|
Spectrum Chemical Mfg Cor
C2362100GM |
100 g | Amber Glass Bottle |
Each for $2,691.59
|
|
||||
|
18609548
|
Spectrum Chemical Mfg Cor
C23621KGBL |
1 kg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 3147-14-6 |
Specifications
| 3147-14-6 | |
| ACS | |
| Brown to Brown/Red | |
| Calmagite |
| 1 | |
| Amber Glass Bottle | |
| 5 g |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.