missing translation for 'onlineSavingsMsg'
Learn More

Calix[8]arene, 97%

Catalog No. p-7025930
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2166603 1 g
AAB2166606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2166603 Supplier Thermo Scientific Chemicals Supplier No. B2166603
Only null left

CAS: 82452-93-5 | C56H48O8 | 848.992 g/mol

It is used in the study on the chromatographic behavior of water-soluble vitamins on p-tert-butyl-calix [8] arene-bonded silica gel stationary phase by HPLC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the study on the chromatographic behavior of water-soluble vitamins on p-tert-butyl-calix [8] arene-bonded silica gel stationary phase by HPLC.

Solubility
Sparingly soluble in water (5.8E-6 g/L) (25°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 82452-93-5
Molecular Formula C56H48O8
Molecular Weight (g/mol) 848.992
MDL Number MFCD00143914
InChI Key HDPRHRZFFPXZIL-UHFFFAOYSA-N
Synonym calix 8 arene, calix 8 arene;, p-h-calix 8 arene, octahydroxycalix 8 arene, para-h-calix 8 arene;, calix 8 arene, technical ch, 49,50,51,52,53,54,55,56-octahydroxycalix 8 arene, 49,50,51,52,53,54,55,56-octahydroxycalix 8 arene;, 49,50,51,52,53,54,55,56-octahydroxycalix 8-arene;, 49,50,51,52,53,54,55,56-octakis hydroxy calix 8 arene;
PubChem CID 2724886
SMILES C1C2=C(C(=CC=C2)CC3=C(C(=CC=C3)CC4=C(C(=CC=C4)CC5=C(C(=CC=C5)CC6=CC=CC(=C6O)CC7=CC=CC(=C7O)CC8=CC=CC(=C8O)CC9=CC=CC1=C9O)O)O)O)O

Specifications

Melting Point >450°C
Quantity 1 g
Solubility Information Sparingly soluble in water (5.8E-6g/L) (25°C).
Formula Weight 848.99
Percent Purity 97%
Chemical Name or Material Calix[8]arene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only