missing translation for 'onlineSavingsMsg'
Learn More

Calix[6]arene 95.0+%, TCI America™

Catalog No. p-7130683
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07131G 1 g
H07135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07131G Supplier TCI America Supplier No. H07131G
Only null left

Chemical Identifiers

CAS 96107-95-8
Molecular Formula C42H36O6
Molecular Weight (g/mol) 636.744
MDL Number MFCD00143083
InChI Key JLSWUKWFQCVKCL-UHFFFAOYSA-N
Synonym calix 6 arene, hexahydroxycalix 6 arene
PubChem CID 2724885
SMILES C1C2=C(C(=CC=C2)CC3=C(C(=CC=C3)CC4=C(C(=CC=C4)CC5=CC=CC(=C5O)CC6=CC=CC(=C6O)CC7=CC=CC1=C7O)O)O)O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 636.74
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Calix[6]arene