missing translation for 'onlineSavingsMsg'
Learn More

Calix[6]arene, 98%

Catalog No. p-7025885
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2160803 1 g
AAB2160806 5 g
AAB2160814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2160803 Supplier Thermo Scientific Chemicals Supplier No. B2160803
Only null left

CAS: 96107-95-8 | C42H36O6 | 636.744 g/mol

It is used as a starting material for preparing functionalized calixarenes. It finds its uses as a pharmaceutical intermediate. It is also used in complexation and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a starting material for preparing functionalized calixarenes. It finds its uses as a pharmaceutical intermediate. It is also used in complexation and organic synthesis.

Solubility
0.180g/100g of water @ 25°C. Miscible with alcohol, chloroform, ether, carbon disulfide, acetone, oils, carbon tetrachloride and glacial acetic acid.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 96107-95-8
Molecular Formula C42H36O6
Molecular Weight (g/mol) 636.744
MDL Number MFCD00143083
InChI Key JLSWUKWFQCVKCL-UHFFFAOYSA-N
Synonym calix 6 arene, hexahydroxycalix 6 arene
PubChem CID 2724885
SMILES C1C2=C(C(=CC=C2)CC3=C(C(=CC=C3)CC4=C(C(=CC=C4)CC5=CC=CC(=C5O)CC6=CC=CC(=C6O)CC7=CC=CC1=C7O)O)O)O

Specifications

Melting Point >300°C
Quantity 1 g
Solubility Information 0.180g/100g of water @ 25°C. Miscible with alcohol,chloroform,ether,carbon disulfide,acetone,oils,carbon tetrachloride and glacial acetic acid.
Formula Weight 636.74
Percent Purity 98%
Chemical Name or Material Calix[6]arene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only