missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calix[4]arene (contains ca. 8% Chloroform) 98.0+%, TCI America™
Supplier: TCI America C14551G
Specifications
| Calix[4]arene (contains ca. 8% Chloroform) | |
| White | |
| MFCD00143912 | |
| calix 4 arene, tetrahydroxycalix 4 arene, calix 4 arene 1g, calix 4 arene-25,26,27,28-tetrol, 25,26,27,28-tetrahydroxycalix 4 aren, 25,26,27,28-tetrahydroxycalix 4 arene, calix 4 arene-25,26,27,28-tetrol, purum hplc | |
| C1C2=C(C(=CC=C2)CC3=C(C(=CC=C3)CC4=CC=CC(=C4O)CC5=CC=CC1=C5O)O)O | |
| 562409 | |
| ≥98.0% (HPLC) |
| 74568-07-3 | |
| C28H24O4 | |
| 1 g | |
| YPNHVQZZPXPQOS-UHFFFAOYSA-N | |
| 424.496 | |
| 424.50 | |
| Crystalline Powder |
Chemical Identifiers
| 74568-07-3 | |
| 424.496 | |
| YPNHVQZZPXPQOS-UHFFFAOYSA-N | |
| 562409 |
| C28H24O4 | |
| MFCD00143912 | |
| calix 4 arene, tetrahydroxycalix 4 arene, calix 4 arene 1g, calix 4 arene-25,26,27,28-tetrol, 25,26,27,28-tetrahydroxycalix 4 aren, 25,26,27,28-tetrahydroxycalix 4 arene, calix 4 arene-25,26,27,28-tetrol, purum hplc | |
| C1C2=C(C(=CC=C2)CC3=C(C(=CC=C3)CC4=CC=CC(=C4O)CC5=CC=CC1=C5O)O)O |
Safety and Handling
TSCA : No