missing translation for 'onlineSavingsMsg'
Learn More

Calix[4]arene (contains ca. 8% Chloroform) 98.0+%, TCI America™

Catalog No. C1455100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1455100MG 100 mg
C14551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1455100MG Supplier TCI America Supplier No. C1455100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 74568-07-3
Molecular Formula C28H24O4
Molecular Weight (g/mol) 424.496
MDL Number MFCD00143912
InChI Key YPNHVQZZPXPQOS-UHFFFAOYSA-N
Synonym calix 4 arene, tetrahydroxycalix 4 arene, calix 4 arene 1g, calix 4 arene-25,26,27,28-tetrol, 25,26,27,28-tetrahydroxycalix 4 aren, 25,26,27,28-tetrahydroxycalix 4 arene, calix 4 arene-25,26,27,28-tetrol, purum hplc
PubChem CID 562409
SMILES C1C2=C(C(=CC=C2)CC3=C(C(=CC=C3)CC4=CC=CC(=C4O)CC5=CC=CC1=C5O)O)O

Specifications

Color White
Quantity 100 mg
Formula Weight 424.50
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Calix[4]arene (contains ca. 8% Chloroform)