missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Calibrex™ organo 525 Bottle Top Dispensers

Adjustable dispensers include a ground glass plunger that is best suited for organic, non-crystallizing acid and base solutions.

Supplier:  DWK Life Sciences W845004

 View more versions of this product

Catalog No. 03410902


Only null left

Description

Description

  • Permanent fluid path visibility
  • Integrated calibration system
  • Choice of adjustment button with each unit
  • Long lasting performance stability
  • Autoclavable at 121°C / 250°F fully assembled
  • Two year warranty
Specifications

Chemical Identifiers

Bottletop Dispenser
Analog
2 Years
Autoclavable
Calibrex organo 525

Specifications

Bottletop Dispenser
0.5 to 5 mL
Calibrex organo 525
Autoclavable
Analog
2 Years
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions