Learn More
BINDER™ Calibration Certificate, Temperature
Calibration Certificate for temperature, measurement in center of chamber at specified temperature.
$1,532.84 - $6,028.58
Specifications
| Duration | 60 to 120 min. |
|---|---|
| Product Type | Calibration Certificate (Temperature) |
| For Use With (Application) | Temperature Measurement |
| Catalog Number | Mfr. No. | For Use With (Equipment) | Calibration Points | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | For Use With (Equipment) | Calibration Points | Price | Quantity & Availability | ||||
|
08112020
|
BINDER™
80121143 |
BINDER Chambers Series MK | 1 Measurement Point |
Each for $1,532.84
|
|
||||
|
08112022
|
BINDER™
80121560 |
BINDER Chambers Series MK | 9 Measurement Points |
Each for $3,634.53
|
|
||||
|
08112023
|
BINDER™
80121581 |
BINDER Chambers Series MK | 15 to 18 Measurement Points |
Each for $4,606.38
|
|
||||
|
08112024
|
BINDER™
80121602 |
BINDER Chambers Series MK | 27 Measurement Points |
Each for $6,028.58
|
|
||||
Description
- Measurement at specified temperature
- Including certificate
- Various points of measurement
Specifications
| 60 to 120 min. | |
| Calibration Certificate (Temperature) | |
| Temperature Measurement |