Learn More
BINDER™ Calibration Certificate, Temperature
Calibration Certificate for temperature, measurement in center of chamber at specified temperature.
Supplier: BINDER™ 80121581
Description
- Measurement at specified temperature
- Including certificate
- Various points of measurement
Chemical Identifiers
| 60 to 120 min. | |
| Calibration Certificate (Temperature) | |
| Temperature Measurement |
Specifications
| BINDER Chambers Series MK | |
| 15 to 18 Measurement Points | |
| Calibration Certificate (Temperature) |
| Calibration certificate, temperature - temperature measurement incl. certificate, 15- 18 measuring points (at specified temperature) | |
| 60 to 120 min. | |
| Temperature Measurement |