missing translation for 'onlineSavingsMsg'
Learn More

Calconcarboxylic Acid, Solstice

Catalog No. 60045217
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045217 10 g Plastic Bottle
60045219 50 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60045217 Supplier Solstice Supplier No. 2126010G

for complexometry

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 3737-95-9
Molecular Formula C21H14N2O7S
Molecular Weight (g/mol) 438.41
MDL Number MFCD00004078
InChI Key ULIVOAKVRBXKKS-PYCFMQQDSA-N
PubChem CID 5895210
IUPAC Name 3-hydroxy-4-[(2Z)-2-(2-oxo-4-sulfonaphthalen-1-ylidene)hydrazinyl]naphthalene-2-carboxylic acid
SMILES C1=CC=C2C(=C1)C=C(C(=C2NN=C3C4=CC=CC=C4C(=CC3=O)S(=O)(=O)O)O)C(=O)O

Specifications

Melting Point 300°C
UN Number NONH for all modes of transport
Beilstein 727492
Formula Weight 438.41g/mol
Packaging Plastic Bottle
Color Violet
Quantity 10 g
Physical Form Powder
Chemical Name or Material Calconcarboxylic Acid, For complexometry
EINECSNumber 223-117-0