missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Sulfate Hemihydrate, Purified (Plaster of Paris), Reagents
Calcium Sulfate Hemihydrate, Purified (Plaster of Paris), Technical, for general laboratory use. Manufactured in ISO 9001 facility.
$88.40 - $227.73
Chemical Identifiers
| CAS | 10034-76-1 |
|---|---|
| Molecular Formula | Ca2H2O9S2 |
| Molecular Weight (g/mol) | 290.28 |
| InChI Key | ZOMBKNNSYQHRCA-UHFFFAOYSA-J |
| Synonym | Plaster of Paris, Gypsum hemihydrate |
| IUPAC Name | dicalcium hydrate disulfate |
| SMILES | O.[Ca++].[Ca++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-337-328
|
Labchem Inc
C21680002A5 |
2 kg |
Each for $227.73
|
|
|||||
|
01-337-327
|
Labchem Inc
C2168000500A5 |
500 g |
Each for $88.40
|
|
|||||
Chemical Identifiers
| 10034-76-1 | |
| 290.28 | |
| Plaster of Paris, Gypsum hemihydrate | |
| O.[Ca++].[Ca++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
| Ca2H2O9S2 | |
| ZOMBKNNSYQHRCA-UHFFFAOYSA-J | |
| dicalcium hydrate disulfate |
Specifications
| Colorless/White | |
| Solid | |
| Ca2H2O9S2 | |
| ZOMBKNNSYQHRCA-UHFFFAOYSA-J | |
| dicalcium hydrate disulfate | |
| Technical | |
| 2.3 |
| 10034-76-1 | |
| 2 kg | |
| Plaster of Paris, Gypsum hemihydrate | |
| O.[Ca++].[Ca++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
| 290.28 | |
| Poly Bottle | |
| Calcium Sulfate Hemihydrate, Purified (Plaster of Paris) |