accessibility menu, dialog, popup

UserName

H2OCaCaOSOOOOSOOO

CAS RN 10034-76-1

IUPAC Name: dicalcium hydrate disulfate
Synonyms: plaster of paris gipshalbhydrat 2CaSO4.H2O calcium sulphate hemihydrate gypsum hemihydrate calcium sulfate hemihydrate
Molecular Weight (g/mol): 290.28
Molecular Formula: Ca2H2O9S2
InChi Key: ZOMBKNNSYQHRCA-UHFFFAOYSA-J
SMILES: O.[Ca++].[Ca++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Grade 
  • (3)
  • (6)
  • (2)
Molecular Weight (g/mol) 
  • (7)
  • (2)
  • (2)
Physical Form 
  • (1)
  • (3)
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (3)
  • (3)
  • (6)
  • (2)
  • (7)
  • (2)
  • (2)
  • (1)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (5)