missing translation for 'onlineSavingsMsg'
Learn More

Calcium Sulfate Hemihydrate, Purified (Plaster of Paris), Reagents

Catalog No. 01337328
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337328 2 kg
01337327 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337328 Supplier Labchem Inc Supplier No. C21680002A5
Only null left

Calcium Sulfate Hemihydrate, Purified (Plaster of Paris), Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10034-76-1
Molecular Formula Ca2H2O9S2
Molecular Weight (g/mol) 290.28
InChI Key ZOMBKNNSYQHRCA-UHFFFAOYSA-J
Synonym Plaster of Paris, Gypsum hemihydrate
IUPAC Name dicalcium hydrate disulfate
SMILES O.[Ca++].[Ca++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color Colorless/White
Physical Form Solid
Quantity 2 kg
Grade Technical
Packaging Poly Bottle
Specific Gravity 2.3
Chemical Name or Material Calcium Sulfate Hemihydrate, Purified (Plaster of Paris)