missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Sulfate, Dihydrate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
Supplier: Avantor AVR145205
Specifications
| Calcium Sulfate | |
| 10101-41-4 | |
| 1 kg | |
| MFCD00149625 | |
| PASHVRUKOFIRIK-UHFFFAOYSA-L | |
| calcium dihydrate sulfate | |
| 24928 | |
| 172.17 | |
| Poly Bottle |
| Dihydrate | |
| Powder | |
| CaH4O6S | |
| calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone | |
| O.O.[Ca++].[O-]S([O-])(=O)=O | |
| 172.16 | |
| CHEBI:32583 | |
| BAKER ANALYZED™ ACS Reagent | |
| 1L = 2.32kg |
Chemical Identifiers
| 10101-41-4 | |
| 172.16 | |
| PASHVRUKOFIRIK-UHFFFAOYSA-L | |
| 24928 | |
| calcium dihydrate sulfate |
| CaH4O6S | |
| MFCD00149625 | |
| calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone | |
| CHEBI:32583 | |
| O.O.[Ca++].[O-]S([O-])(=O)=O |