missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Sulfate, Dihydrate, NF, 98-101%, Spectrum™ Chemical
CA170, 10101-41-4, CaSO4·2H2O
Supplier: Spectrum Chemical Mfg Cor CA17012KG
Description
Spectrum™ Chemical Calcium Sulfate, Dihydrate, NF is used as an excipient for its filler and diluent properties. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 10101-41-4 | |
| CaH4O6S | |
| 12 kg | |
| O.O.[Ca++].[O-]S([O-])(=O)=O | |
| 172.16 | |
| NF | |
| Poly Pail |
| 100% | |
| MFCD00149625 | |
| PASHVRUKOFIRIK-UHFFFAOYSA-L | |
| calcium dihydrate sulfate | |
| 98 to 101% | |
| 19.0-23% |
Chemical Identifiers
| 10101-41-4 | |
| 172.16 | |
| PASHVRUKOFIRIK-UHFFFAOYSA-L | |
| O.O.[Ca++].[O-]S([O-])(=O)=O |
| CaH4O6S | |
| MFCD00149625 | |
| calcium dihydrate sulfate |