missing translation for 'onlineSavingsMsg'
Learn More

Calcium Sulfate, Dihydrate, NF, 98-101%, Spectrum™ Chemical

Catalog No. 18601399
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601399 500 g Poly Bottle
18601400 2.5 kg Poly Pail
18601401 12 kg Poly Pail
18601402 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601399 Supplier Spectrum Chemical Mfg Cor Supplier No. CA170500GM
Only null left

CA170, 10101-41-4, CaSO4·2H2O

Spectrum™ Chemical Calcium Sulfate, Dihydrate, NF is used as an excipient for its filler and diluent properties. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10101-41-4
Molecular Formula CaH4O6S
Molecular Weight (g/mol) 172.16
MDL Number MFCD00149625
InChI Key PASHVRUKOFIRIK-UHFFFAOYSA-L
IUPAC Name calcium dihydrate sulfate
SMILES O.O.[Ca++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 98 to 101%
Grade NF
Loss on Drying 19.0-23%
Packaging Poly Bottle