missing translation for 'onlineSavingsMsg'
Learn More

Calcium sulfate dihydrate, 99%

Catalog No. p-7050014
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA33301A1 1 kg
AA33301A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA33301A1 Supplier Thermo Scientific Chemicals Supplier No. 033301A1
Only null left

CAS: 10101-41-4 | CaH4O6S | 172.16 g/mol

Used as portland-cement retarder, tile and plaster. It is a source of sulfur and sulfuric acid. Also used in polishing powder and paints, dyeing and calico printing. It is likewise employed in zinc metallurgy. It is employed in drying industrial gases, solids and many organic liquids. In granulated form it is used as soil conditioner. Used for food additives in the soy manufacturing industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10101-41-4
Molecular Formula CaH4O6S
Molecular Weight (g/mol) 172.16
MDL Number MFCD00149625
InChI Key PASHVRUKOFIRIK-UHFFFAOYSA-L
Synonym calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone
PubChem CID 24928
ChEBI CHEBI:32583
IUPAC Name calcium;sulfate;dihydrate
SMILES O.O.[Ca++].[O-]S([O-])(=O)=O

Specifications

Boiling Point 163°C (in - 2H2O)
Melting Point 128°C (in -1.5H2O)
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99%
Merck Index 14,1706
Solubility Information Soluble in water.
Formula Weight 172.17 (136.14 Anhydrous)
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Density 2.32 g/mL
Chemical Name or Material Calcium sulfate dihydrate
Show More Show Less
EINECSNumber 231-900-3
RTECSNumber EW4150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only