missing translation for 'onlineSavingsMsg'
Learn More

Calcium sulfate dihydrate, 98+%, extra pure

Catalog No. p-4380025
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC446380010 1 kg Plastic bottle
AC446380050 5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446380010 Supplier Thermo Scientific Chemicals Supplier No. 446380010
Only null left

CAS: 10101-41-4 | CaH4O6S | 172.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10101-41-4
Molecular Formula CaH4O6S
Molecular Weight (g/mol) 172.16
MDL Number MFCD00149625
InChI Key PASHVRUKOFIRIK-UHFFFAOYSA-L
Synonym calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone
PubChem CID 24928
ChEBI CHEBI:32583
IUPAC Name calcium;sulfate;dihydrate
SMILES O.O.[Ca++].[O-]S([O-])(=O)=O

Specifications

Quantity 1 kg
Assay Percent Range 98% min. (Complexometry)
Linear Formula CaSO4·2H2O
Solubility Information Solubility in water: 2g/L (20°C). Other solubilities: slightly soluble in glycerol, practically insoluble in most common organic, solvents
Formula Weight 172.17
Percent Purity ≥98%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Calcium sulfate dihydrate
Hazard Category

RUO – Research Use Only