missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Sulfate, Anhydrous, FCC, 98%, Spectrum™ Chemical
CA171, 7778-18-9, CaSO4
$390.18 - $5455.13
Chemical Identifiers
| CAS | 7778-18-9 |
|---|---|
| Molecular Formula | CaO4S |
| Molecular Weight (g/mol) | 136.13 |
| MDL Number | MFCD00010912 |
| InChI Key | OSGAYBCDTDRGGQ-UHFFFAOYSA-L |
| IUPAC Name | calcium sulfate |
| SMILES | [Ca++].[O-]S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18601391
|
Spectrum Chemical Mfg Cor
CA171500GM |
500 g | Poly Bottle |
Each for $390.18
|
|
||||
|
18601392
|
Spectrum Chemical Mfg Cor
CA17125KG |
2.5 kg | Poly Pail |
Each for $1,197.63
|
|
||||
|
18601393
|
Spectrum Chemical Mfg Cor
CA17112KG |
12 kg | Poly Pail |
Each for $3,113.53
|
|
||||
|
18601394
|
Spectrum Chemical Mfg Cor
CA17145KGBL |
45 kg | Fiber Drum |
Each for $5,455.13
|
|
||||