Learn More
Calcium sulfate, anhydrous, 99%
CAS: 7778-18-9 | CaO4S | 136.13 g/mol
Supplier: Thermo Scientific Chemicals 040144A1
Description
Used as Portland-cement retarder; tile and plaster; It is the source of sulfur and sulfuric acid; Also used in polishing powder and paints, dyeing and calico printing. It is likewise employed in zinc metallurgy. It is employed in drying industrial gases, solids and many organic liquids. In granulated form it is used as soil conditioner. Used for food additives in the soy manufacturing industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Calcium sulfate, anhydrous | |
| 7778-18-9 | |
| Powder | |
| 99% | |
| MFCD00010912 | |
| Soluble in water,slightly soluble in glycerol. | |
| [Ca++].[O-]S([O-])(=O)=O | |
| 136.13 | |
| CHEBI:31346 | |
| Hygroscopic | |
| 2.96 g/cm3 at 20°C |
| 1450°C | |
| White | |
| 1 kg | |
| CaO4S | |
| calcium sulfate, drierite, gypsum, sulfuric acid, calcium salt 1:1, karstenite, calcium sulphate, anhydrous gypsum, plaster of paris, anhydrous calcium sulfate, calcium sulfate, anhydrous | |
| OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
| calcium;sulfate | |
| 24497 | |
| 136.14 | |
| White powder |
Chemical Identifiers
| 7778-18-9 | |
| 136.13 | |
| OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
| 24497 | |
| calcium;sulfate |
| CaO4S | |
| MFCD00010912 | |
| calcium sulfate, drierite, gypsum, sulfuric acid, calcium salt 1:1, karstenite, calcium sulphate, anhydrous gypsum, plaster of paris, anhydrous calcium sulfate, calcium sulfate, anhydrous | |
| CHEBI:31346 | |
| [Ca++].[O-]S([O-])(=O)=O |
Safety and Handling
H315-H319 : Causes skin irritation. Causes serious eye irritation.
EINECSNumber : 231-900-3
RTECSNumber : WS6920000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only