missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium sulfate, 99%, pure, anhydrous
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
Specifications
Specifications
| Melting Point | 1450.0°C |
| Color | White |
| Physical Form | Fine Crystalline Powder |
| Quantity | 500 g |
| Packaging | Plastic bottle |
| Assay Percent Range | 98.5% min |
| Linear Formula | CaSO4 |
| Fieser | 01,107 |
| Merck Index | 15,1708 |
| Solubility Information | Solubility in water: slightly soluble. Other solubilities: slightly soluble in glycerol,practically insoluble in most common organic,solvents |
| Show More |
Safety and Handling
| Hazard Category | |
| EINECSNumber | 231-900-3 |
RUO – Research Use Only