missing translation for 'onlineSavingsMsg'
Learn More

Calcium Stearate, FCC, 9-10.5%, Spectrum™ Chemical

Catalog No. 18601381
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601381 500 g Poly Pail
18601382 2.5 kg Poly Pail
18601383 12 kg Fiber Drum
18601384 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601381 Supplier Spectrum Chemical Mfg Cor Supplier No. C1131500GM

C1131, 1592-23-0

Spectrum™ Chemical Calcium Stearate, FCC , also known as E470, is used as a flow agent in powders. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1592-23-0
Molecular Formula C36H70CaO4
Molecular Weight (g/mol) 607.03
InChI Key CJZGTCYPCWQAJB-UHFFFAOYSA-L
IUPAC Name calcium dioctadecanoate
SMILES [Ca++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 9 to 10.5%
Grade FCC
Loss on Drying 4%
Packaging Poly Pail