missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium stearate, 5.9 to 7.1% (Ca), powder, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 145°C to 160°C |
| Physical Form | Powder |
| Quantity | 5 kg |
| Packaging | Plastic drum |
| Merck Index | 15,1705 |
| Solubility Information | Solubility in water: insoluble |
| Formula Weight | 607.02 |
| Percent Purity | 6.4 to 7.4% (Ca) |
| Chemical Name or Material | Calcium stearate, 6.4 to 7.4% (Ca) |
Safety and Handling
| EINECSNumber | 216-472-8 |
RUO – Research Use Only