missing translation for 'onlineSavingsMsg'
Learn More

Calcium stearate, 5.9 to 7.1% (Ca), powder, Thermo Scientific Chemicals

Catalog No. AC449720010
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC449720010 1 kg
AC449720050 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC449720010 Supplier Thermo Scientific Chemicals Supplier No. 449720010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1592-23-0
Molecular Formula C36H70CaO4
Molecular Weight (g/mol) 607.02
MDL Number MFCD00036390
InChI Key CJZGTCYPCWQAJB-UHFFFAOYSA-L
Synonym calcium stearate, calcium octadecanoate, flexichem, calcium distearate, octadecanoic acid, calcium salt, aquacal, calstar, flexichem cs, stavinor 30, stearates
PubChem CID 15324
IUPAC Name calcium;octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Ca+2]

Specifications

Melting Point 145°C to 160°C
Physical Form Powder
Quantity 1 kg
Merck Index 15,1705
Solubility Information Solubility in water: insoluble
Formula Weight 607.02
Percent Purity 6.4 to 7.4% (Ca)
Packaging Plastic Bottle
Chemical Name or Material Calcium stearate, 6.4 to 7.4% (Ca)
EINECSNumber 216-472-8

RUO – Research Use Only