missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium standard solution, for AAS, 1 mg/ml Ca in 2 to 5% HNO3
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for atomic adsorption spectroscopy
Specifications
Specifications
| Chemical Name or Material | Calcium standard solution |
| Packaging | Plastic bottle |
| Name Note | 1 mg/ml Ca in 0.5N HNO3 |
| CAS | 7440-70-2, 7697-37-2, 7732-18-5 |
| Physical Form | Solution |
| Molecular Formula | Ca |
| Concentration | 997 to 1003 μg/mL of Ca(2+) in 0.5N HNO3 (20°C) |
| MDL Number | MFCD00010897 MFCD00085314 |
| Solubility | Soluble in water |
| InChI Key | OYPRJOBELJOOCE-UHFFFAOYSA-N |
| Show More |
RUO – Research Use Only