missing translation for 'onlineSavingsMsg'
Learn More

Calcium Standard, For AAS, 1,000 mg/L Ca in Nitric Acid, MilliporeSigma™ Supelco™

Catalog No. 111020413
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020413 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020413 Supplier Merck Emd Millipore Supplier No. 69349250ML

For a complete product listing of our TraceCERT range of CRMs for ICP and AAS, technical information and example certificates please visit our inorganic TraceCERT website

Specifications

Product Line TraceCERT
Concentration 1000 mg/L Ca in nitric acid
Quantity 250 mL
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC