missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Calcium Sensor Dye eFluor™ 514
Description
- Calcium binding affinity: Kd = 232 nM
- Molecular Weight: ≈1100 d
- Peak Excitation: 490nm
- Peak Emission: 514nm
- Reconstitute in high-quality, freshly opened DMSO
- Recommended concentration: 2 to 5 mM
- Once reconstituted, protect from light and store dessicated at −20°C; avoid freeze-thawing
Specifications
Specifications
| Detection Method | Fluorescence |
| For Use With (Application) | Calcium Assay |
| For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
| Product Type | Dye |
| Product Line | eBioscience |
| Quantity | 10 x 50μg |
| Emission Wavelength | 490/514 |
For Research Use Only. Not for use in diagnostic procedures.