Learn More
Calcium Saccharate, Tetrahydrate, USP, 98.5-102%, Spectrum™ Chemical
C1830, 5793-89-5, C6H8CaO8·4H2O
Supplier:  Spectrum Chemical Mfg Cor C183012KGBL
 View more versions of this product
Catalog No. 18601369
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
Spectrum™ Chemical Calcium Saccharate, Tetrahydrate, USP is used as an antacid, as an antidote in carbolic acid poisoning, and as a stabilizer for calcium gluconate solution for parenteral administration. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
5793-89-5
12 kg
ZQWFSIZRQANUDA-FARGFPGJSA-L
calcium (2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanedioate tetrahydrate
98.5 to 102%
Poly Pail
100%
C6H16CaO12
O.O.O.O.[Ca++].O[C@@H]([C@@H](O)[C@@H](O)C([O-])=O)[C@H](O)C([O-])=O
320.26
USP
Chemical Identifiers
5793-89-5
320.26
calcium (2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanedioate tetrahydrate
C6H16CaO12
ZQWFSIZRQANUDA-FARGFPGJSA-L
O.O.O.O.[Ca++].O[C@@H]([C@@H](O)[C@@H](O)C([O-])=O)[C@H](O)C([O-])=O
Product Suggestions
Product Suggestions