missing translation for 'onlineSavingsMsg'
Learn More

Calcium Saccharate, Tetrahydrate, USP, 98.5-102%, Spectrum™ Chemical

Catalog No. p-7157197
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
12 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601366 25 g Amber Glass Bottle
18601367 100 g Amber Glass Bottle
18601368 500 g Amber Glass Bottle
18601369 12 kg Poly Pail
18601370 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18601366 Supplier Spectrum Chemical Mfg Cor Supplier No. C183025GM
Only null left

C1830, 5793-89-5, C6H8CaO8·4H2O

Spectrum™ Chemical Calcium Saccharate, Tetrahydrate, USP is used as an antacid, as an antidote in carbolic acid poisoning, and as a stabilizer for calcium gluconate solution for parenteral administration. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 5793-89-5
Molecular Formula C6H16CaO12
Molecular Weight (g/mol) 320.26
InChI Key ZQWFSIZRQANUDA-FARGFPGJSA-L
IUPAC Name calcium (2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanedioate tetrahydrate
SMILES O.O.O.O.[Ca++].O[C@@H]([C@@H](O)[C@@H](O)C([O-])=O)[C@H](O)C([O-])=O

Specifications

CAS Max % 100%
Quantity 25 g
Percent Purity 98.5 to 102%
Grade USP
Packaging Amber Glass Bottle